missing translation for 'onlineSavingsMsg'
Learn More

PHA 767491 Dihydrochloride Salt, TRC

Product Code. 30422199
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422199 25 mg 25mg
30418542 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422199 Supplier Toronto Research Chemicals Supplier No. TRCP29445025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,5,6,7-Tetrahydro-2-(4-pyridinyl)-4H-pyrrolo[3,2-c]pyridin-4-one Dihydrochloride
Analyte or Component Names 1,5,6,7-Tetrahydro-2-(4-pyridinyl)-4H-pyrrolo[3,2-c]pyridin-4-one Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H11 N3 O . 2 Cl H
Synonym PHA 767491 Dihydrochloride Salt, PF-02334471
InChI Formula InChI=1S/C12H11N3O.2ClH/c16-12-9-7-11(8-1-4-13-5-2-8)15-10(9)3-6-14-12;;/h1-2,4-5,7,15H,3,6H2,(H,14,16);2*1H
Solution Type Neat
SMILES Cl.Cl.O=C1NCCc2[nH]c(cc12)c3ccncc3
IUPAC Name 2-(4-pyridyl)-1,5,6,7-tetrahydropyrrolo[3,2-c]pyridin-4-one;hydrochloride
Industry Type Forensics
Molecular Weight (g/mol) 286.16
Quantity 25 mg
Formula Weight 285.0436
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.