missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ pH Combination Electrode for Tris Buffer
Click to view available options
Connector Types:
BNC
Unit Size:
Each
Description
- Refillable, combination electrode
- With Calomel references
- BNC connector
Specifications
Specifications
| Connector Types | BNC |
| Diameter (Metric) Cap | 9.5 mm |
| Junction | Ceramic |
| Length (Metric) Cable | 1 m |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction