missing translation for 'onlineSavingsMsg'
Learn More

PH-797804, MedChemExpress

Product Code. 16496647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496647 5 mg 5mg
16490287 50 mg 50mg
16474956 100 mg 100mg
16420986 200 mg 200mg
16413938 10 mg 10mg
16462916 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496647 Supplier Medchem Express Supplier No. HY10403

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PH-797804 is a ATP-competitive, selective p38α/p38β inhibitor (IC50=26 nM and Ki=5.8 nM for p38α; Ki=40 nM for p38β) and does not inhibit JNK2.

PH-797804 is a ATP-competitive, selective p38α/p38β inhibitor (IC50=26 nM and Ki=5.8 nM for p38α

Specifications

Chemical Name or Material PH-797804
CAS 586379-66-0
Color White
Purity Grade Notes Research
Molecular Formula C22H19BrF2N2O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (104.76 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(N2C(C)=CC(OCC3=CC=C(C=C3F)F)=C(C2=O)Br)=C1)C)NC
Molecular Weight (g/mol) 477.3
Formula Weight 477.3
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.