missing translation for 'onlineSavingsMsg'
Learn More

PGP9.5 / UchL1 (pan-Neuronal Marker) Mouse, Clone: rUCHL1/775, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 7345-MSM5-P1

 View more versions of this product

Product Code. 15991048

  • ONLINE EXCLUSIVE PRICE
    554.40 EUR
    478.00 EUR / 100µg
    Save 76.40 EUR
    13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

PGP9.5 / UchL1 (pan-Neuronal Marker)
Cytoplasmic. Endoplasmic Reticulum membrane
rUCHL1/775
Unconjugated
Mouse
20-30kDa
Protein A/G purified from Bioreactor concentrate.
RUO
7345
IgG1 κ
Western Blot, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human UCHL1 protein
Cerebellum
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.