Learn More
Cole-Parmer™ PFA Threaded Reducing Bushing
Click to view available options
Thread Size:
1/2 in. NPTM x 1/4 in. NPTF
1/2 in. NPTM x 3/8 in. NPTF
1/4 in. NPTM x 1/8 in. NPTF
3/4 in. NPTM x 1/2 in. NPTF
3/8 in. NPTM x 1/4 in. NPTF
3/8 in. NPTM x 1/8 in. NPTF
Unit Size:
Each
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
| Description | NPT(M) x NPT(F) Connection |
| Autoclavable | Autoclavable |
| Material | PFA |
| Outlet Port | 3/8 in. |
| Port Size | 1/2 in. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Thread Size | 1/2 in. NPTM x 3/8 in. NPTF |
| Color | Translucent |
| Product Type | Threaded Reducing Bushing |