missing translation for 'onlineSavingsMsg'
Learn More

PF-CBP1 hydrochloride, MedChemExpress

Product Code. p-200098650
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488008

Brand: Medchem Express HY19999A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-CBP1 hydrochloride is a highly selective inhibitor of the CREB binding protein bromodomain (CBP BRD).

PF-CBP1 inhibits CREBBP and EP300 bromodomains with IC50 of 125 nM and 363 nM respectively. PF-CBP1 hydrochloride reduces LPS-induced inflammatory cytokines expression (IL-1β, IL-6 and IFN-β) in primary macrophages. PF-CBP1 hydrochloride also downregulates RGS4 expression cortical neurons and can be used for the research of neurological disorders, including epilepsy and parkinson's disease, et al.

Specifications

Chemical Name or Material PF-CBP1 hydrochloride
CAS 2070014-93-4
Color White
Purity Grade Notes Research
Molecular Formula C29H37ClN4O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (190.45 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.90 mM; Need ultrasonic)
SMILES CCCOC1=CC=C(CCC2=NC3=CC(C4=C(C)ON=C4C)=CC=C3N2CCN5CCOCC5)C=C1.Cl[H]
Molecular Weight (g/mol) 525.08
Formula Weight 525.08
Percent Purity 95.95%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.