missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-CBP1 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
PF-CBP1 inhibits CREBBP and EP300 bromodomains with IC50 of 125 nM and 363 nM respectively. PF-CBP1 hydrochloride reduces LPS-induced inflammatory cytokines expression (IL-1β, IL-6 and IFN-β) in primary macrophages. PF-CBP1 hydrochloride also downregulates RGS4 expression cortical neurons and can be used for the research of neurological disorders, including epilepsy and parkinson's disease, et al.
Specifications
Specifications
| Chemical Name or Material | PF-CBP1 hydrochloride |
| CAS | 2070014-93-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H37ClN4O3 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 100 mg/mL (190.45 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.90 mM; Need ultrasonic) |
| SMILES | CCCOC1=CC=C(CCC2=NC3=CC(C4=C(C)ON=C4C)=CC=C3N2CCN5CCOCC5)C=C1.Cl[H] |
| Molecular Weight (g/mol) | 525.08 |
| Formula Weight | 525.08 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction