Learn More
PF-9366, MedChemExpress
Product Code. 16455116
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455116 100 mg 100mg
16442976 25 mg 25mg
16424158 10 mg 10mg
16420507 50 mg 50mg
16406867 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16455116 Supplier Medchem Express Supplier No. HY107778
MedChemExpress PF-9366 is a human methionine adenosyltransferase 2A (Mat2A) inhibitor, with an IC50 of 420 nM and a Kd of 170 nM.
PF-9366 is a human methionine adenosyltransferase 2A (Mat2A) inhibitor, with an IC50 of 420 nM and a Kd of 170 nM.
Specifications
Chemical Name or Material PF-9366
CAS 72882-78-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H19ClN4
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (71.26 mM; Need ultrasonic)
SMILES CN(C)CCC1=NN=C2N1C3=CC=C(Cl)C=C3C(C4=CC=CC=C4)=C2
Molecular Weight (g/mol) 350.84
Formula Weight 350.84
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death