missing translation for 'onlineSavingsMsg'
Learn More

PF-8380, MedChemExpress

Product Code. 16495658
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495658 10 mg 10mg
16461827 50 mg 50mg
16448357 5 mg 5mg
16436176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495658 Supplier Medchem Express Supplier No. HY13344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-8380 is a potent autotaxin inhibitor with an IC50 of 2.8 nM in isolated enzyme assay and 101 nM in human whole blood.

PF-8380 is a potent autotaxin inhibitor with an IC50 of 2.8 nM in isolated enzyme assay and 101 nM in human whole blood.

Specifications

Chemical Name or Material PF-8380
CAS 1144035-53-9
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2N3O5
Quantity 10 mg
Solubility Information DMSO : 6.67 mg/mL (13.94 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES O=C(N1CCN(CC1)CCC(C2=CC(O3)=C(C=C2)NC3=O)=O)OCC4=CC(Cl)=CC(Cl)=C4
Molecular Weight (g/mol) 478.33
Formula Weight 478.33
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.