missing translation for 'onlineSavingsMsg'
Learn More

PF-4136309, MedChemExpress

Product Code. 16408287
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16408287

Brand: Medchem Express HY13245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC50s of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2.

PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC50s of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2.

Specifications

Chemical Name or Material PF-4136309
CAS 1341224-83-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31F3N6O3
Quantity 5 mg
Synonym INCB8761
Solubility Information Ethanol : 133.33 mg/mL (234.49 mM; Need ultrasonic) ∣DMSO : ≥ 34 mg/mL (59.80 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O[C@]1(CC[C@](CC1)([H])N[C@@H]2CN(CC2)C(CNC(C3=CC=CC(C(F)(F)F)=C3)=O)=O)C4=NC=C(C=C4)C5=NC=CC=N5
Molecular Weight (g/mol) 568.59
Formula Weight 568.59
Percent Purity 96.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.