Learn More
Tocris Bioscience™ PF 3758309 Dihydrochloride, Tocris Bioscience™
GREENER_CHOICE
Potent PAK4 inhibitor; orally available
Brand:  Tocris Bioscience™ 6005/50
Encompass
Product Code. 15706815
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PF 3758309 dihydrochloride is a potent PAK4 inhibitor (IC50 = 1.3 nM; Kd = 2.7 nM). Inhibits proliferation and anchorage-independent growth of a panel of tumor cell lines in vitro. Suppresses growth of human HCT116 and A549 tumor cell xenografts in mice. Orally available.
Specifications
Specifications
PF 3758309 Dihydrochloride
50 mg
Solid
Bioactive Small Molecule
N-[(1S)-2-(Dimethylamino)-1-phenylethyl]-4,6-dihydro-6,6-dimethyl-3-[(2-methylthieno[3,2-d]pyrimidin-4-yl)amino]pyrrolo[3,4-c]pyrazole-5(1H)-carboxamide dihydrochloride
Soluble to 100mM in water and to 100mM in DMSO
Off-white
Certificates
Promotions
Promotions
Product Title
Select an issue
For Research Use Only.