missing translation for 'onlineSavingsMsg'
Learn More

PF-06869206, MedChemExpress

Product Code. 16470857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470857

Brand: Medchem Express HY112065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06869206 is an orally bioavailable selective inhibitor of the sodium-phosphate cotransporter NaPi2a (SLC34A1) with an IC50 of 380 nM.

PF-06869206 is an orally bioavailable selective inhibitor of the sodium-phosphate cotransporter NaPi2a (SLC34A1) with an IC50 of 380 nM.

Specifications

Chemical Name or Material PF-06869206
CAS 2227425-05-8
Color Light Nude
Purity Grade Notes Research
Molecular Formula C15H14ClF3N4O2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (266.84 mM; Need ultrasonic)
SMILES ClC1=C(C)NC2=C(N3C[C@@H](CO)OCC3)C(C#N)=C(C(F)(F)F)N=C21
Molecular Weight (g/mol) 374.75
Formula Weight 374.75
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.