missing translation for 'onlineSavingsMsg'
Learn More

PEX1, Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16162615
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16162615 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16162615 Supplier Abnova Supplier No. H00005189B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MWGSDRLAGAGGGGAAVTVAFTNARDCFLHLPRRLVAQLHLLQNQAIEVVWSHQPAFLSWVEGRHFSDQGENVAEINRQVGQKLGLSNGGQVFLKPCSHVVSCQQVEVEPLSADDWEILELHAVSLEQHLLDQIRIVFPKAIFPVWVDQQTYIFIQIVALIPAASYGRLETDTKLLIQPKTRRAKENTFSKADAEYKKLHSYGRDQKGMMKELQTKQLQSNTVGITESNENESEIPVDSSSVASLWTMIGSIFSFQSEKKQETSWGLTEINAFKNMQSKVVPLDNIFRVCKSQPPSIYNASATSVFHKHCAIHVFPWDQEYFDVEPSFTVTYGKLVKLLSPKQQQSKTKQNVLSPEKEKQMSEPLDQKKIRSDHNEEDEKACVLQVVWNGLEELNNAIKYTKNVEVLHLGKVWIPDDLRKRLNIEMHAVVRITPVEVTPKIPRSLKLQPRENLPKDISEEDIKTVFYSWLQQSTTTMLPLVISEEEFIKLETKDGLKEFSLSIVHSWEKEKDKNIFLLSPNLLQKTTIQVLLDPMVKEENSEEIDFILPFLKLSSLGGVNSLGVSSLEHITHSLLGRPLSRQLMSLVAGLRNGALLLTGGKGSGKSTLAKAICKEAFDKLDAHVERVDCKALRGKRLENIQKTLEVAFSEAVWMQPSVVLLDDLDLIAGLPAVPEHEHSPDAVQSQRLAHALNDMIKEFISMGSLVALIATSQSQQSLHPLLVSAQGVHIFQCVQHIQPPNQEQRCEILCNVIKNKLDCDINKFTDLDLQHVAKETGGFVARDFTVLVDRAIHSRLSRQSISTREKLVLTTLDFQKALRGFLPASLRSVNLHKPRDLGWDKIGGLHEVRQILMDTIQLPAKYPELFANLPIRQRTGILLYGPPGTGKTLLAGVIARESRMNFISVKGPELLSKYIGASEQAVRDIFIRAQAAKPCILFFDEFESIAPRRGHDNTGVTDRVVNQLLTQLDGVEGLQGVYVLAATSRPDLIDPALLRPGRLDKCVYCPPPDQVSRLEILNVLSDSLPLADDVDLQHVASVTDSFTGADLKALLYNAQLEALHGMLLSSGLQDGSSSSDSDLSLSSMVFLNHSSGSDDSAGDGECGLDQSLVSLEMSEILPDESKFNMYRLYFGSSYESELGNGTSSDLSSQCLSAPSSMTQDLPGVPGKDQLFSQPPVLRTASQEGCQELTQEQRDQLRADISIIKGRYRSQSGEDESMNQPGPIKTRLAISQSHLMTALGHTRPSISEDDWKNFAELYESFQNPKRRKNQSGTMFRPGQKVTLA

Specifications

Antigen PEX1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene peroxisomal biogenesis factor 1
Gene Accession No. NM_000466.2
Gene Alias ZWS1
Gene Symbols PEX1
Host Species Mouse
Immunogen PEX1 (NP_000457.1, 1 a.a. ∼ 1283 a.a) full-length human protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5189
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.