Learn More
Pesticide Mix 4, SPEX CertiPrep™
Product Code. 16310488 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16310488 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16310488 Supplier SPEX Certiprep Supplier No. SPXPR4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 4 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
  • Pesticide Mix 4 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Specifications
Chemical Name or Material Epn
Dichlorvos
Edifenphos
Ethion
Fenitrothion
Ethyl parathion
Methyl parathion
Packaging Glass Ampule
Assay Percent Range 99.1
98.8
97.5
99.5
98
99
99.6
99.5
99.5
99.2
99.2
99.5
98
99.5
98
Molecular Formula C14H14NO4PS
C4H7Cl2O4P
C14H15O2PS2
C9H22O4P2S4
C9H12NO5PS
C10H14NO5PS
C8H10NO5PS
C4H10NO3PS
C8H19O2PS3
C10H15O3PS2
C12H21N2O3PS
C9H11Cl3NO3PS
C12H4Cl2F6N4OS
C12H6F2N2O2
C8Cl4N2
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 323.304 g/mol
220.976 g/mol
310.371 g/mol
384.476 g/mol
277.234 g/mol
291.261 g/mol
263.207 g/mol
183.166 g/mol
274.404 g/mol
278.328 g/mol
304.345 g/mol
350.59 g/mol
437.148 g/mol
248.185 g/mol
265.911 g/mol
compliance-icons compliance-icons
Product Identifier
  • Pesticide Mix 4
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetonitrile