Learn More
Pesticide Mix 3, SPEX CertiPrep™
Product Code. 16300488 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16300488 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16300488 Supplier SPEX Certiprep Supplier No. SPXPR3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 3 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
  • Pesticide Mix 3 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Warranty
1 year
Specifications
Chemical Name or Material Propargite (omite)
Carbaryl
Myclobutanil (systhane)
Dimethomorph
Etoxazole
Spirodiclo
Packaging Glass Ampule
Assay Percent Range 97.4
99.9
98.6
98.4
98.9
99.5
99.6
99.5
99.5
98.5
99
96
98.9
99
99.4
Molecular Formula C19H26O4S
C12H11NO2
C15H17ClN4
C21H22ClNO4
C21H23F2NO2
C21H24Cl2O4
C8H10ClN5O3S
C9H6F3N3O
C25H28O3
C7H17O2PS3
C2H8NO2PS
C11H15BrClO3PS
C7H14NO5P
C12H17O4PS2
C11H20N3O3PS
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 350.472 g/mol
201.221 g/mol
288.775 g/mol
387.8567 g/mol
359.410 g/mol
411.319 g/mol
291.715 g/mol
229.159 g/mol
376.488 g/mol
260.377 g/mol
141.001328 g/mol
373.631 g/mol
223.1635 g/mol
320.3647 g/mol
305.334 g/m
compliance-icons compliance-icons
Product Identifier
  • Pesticide Mix 3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetonitrile