Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
Product Code. 12772916 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
Unit Size:
Pack of 4800
Pack of 960
Product Code. 12772916
Brand: Axygen™ RRP50CBKR
Only null left
Add to basket
Add to basket
Volume ranges from 20 to 200μL
  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Rack Color Green
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Liquid Level Sensing, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head