Learn More
Contec™ Trigger Sprayer for PeridoxRTU™ Sporicidal Disinfectant/Cleaner
Shop All Contec Products
Click to view available options
Volume (Metric):
0.9 L
Unit Size:
Pack of 6
Description
- Foaming trigger sprayer
- For daily cleaning and disinfecting tasks, such as wiping surfaces and mopping floors.
- Ideal for use in pharmaceutical, biopharmaceutical and medical device facilities.
Specifications
Specifications
| Product Type | Foaming Trigger Sprayer |
| Color-coded | No |
| Form | Foam |
| Scent | Original |
| Sterility | Non-sterile |
| For Use With (Application) | Daily cleaning and disinfecting tasks |
| For Use With (Equipment) | For use with 32 oz. Bottle of PeridoxRTU (CR85335IR) |
| Size | 0.9 L |
| Volume (Metric) | 0.9 L |