Learn More
Perchloric Acid, 0.1M, Chem Lab™
Product Code. 12628533
Change view
Click to view available options
Quantity:
1 L
Grade:
Laboratory
Reagent Acc. Ph. Eur. 9(4.2.2)
Unit Size:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Grade unitSize
12628533 1 L Reagent Acc. Ph. Eur. 9(4.2.2) 1 L
11973263 1 L Laboratory 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12628533 Supplier Chem-Lab Supplier No. CL07.1608.1000
Specifications
pH <2
Color Colorless
Density 1.05g/mL
Odor Acetic Acid
Packaging 1L
Quantity 1 L
Physical Form Liquid
Grade Reagent Acc. Ph. Eur. 9(4.2.2)
compliance-icons compliance-icons
Product Identifier
  • Perchloric acid 0.1 mol/l in Acetic acid 100%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Perchloric acid