missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEN2 Recombinant Rabbit Monoclonal Antibody (6J5O4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538177
This item is not returnable.
View return policy
Description
Positive Samples: 293T, A-549, SH-SY5Y, Mouse kidney, Rat lung Immunogen sequence: MNLERVSNEE KLNLCRKYYL GGFAFLPFLW LVNIFWFFRE AFLVPAYTEQ SQIKGYVWRS AVGFLFWVIV LTSWITIFQI YRPRWGALGD YLSFTIPLGT P.
Presenilins, which are components of the gamma-secretase protein complex, are required for intramembranous processing of some type I transmembrane proteins, such as the Notch proteins and the beta-amyloid precursor protein. Signaling by Notch receptors mediates a wide range of developmental cell fates. Processing of the beta-amyloid precursor protein generates neurotoxic amyloid beta peptides, the major component of senile plaques associated with Alzheimer's disease. This gene encodes a protein that is required for Notch pathway signaling, and for the activity and accumulation of gamma-secretase.
Specifications
| PEN2 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6QI68, Q9CQR7, Q9NZ42 | |
| PSENEN | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6J5O4 | |
| Unconjugated | |
| PSENEN | |
| 1700023M09Rik; gamma-secretase subunit PEN-2; hematopoietic stem/progenitor cells protein MDS033; liver regeneration-related protein LRRGT00140; LRRGT00140 mRNA; MDS033; MSTP064; Pen2; Pen-2; presenilin enhancer 2; presenilin enhancer 2 homolog; presenilin enhancer 2 homolog (C. elegans); presenilin enhancer gamma secretase subunit; presenilin enhancer gamma-secretase subunit; presenilin enhancer protein 2; presenilin enhancer, gamma-secretase subunit; Psenen; RGD1312037 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 292788, 55851, 66340 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction