missing translation for 'onlineSavingsMsg'
Learn More

Pefloxacinium, TRC

Product Code. 17943825
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943825 1 g 1g
17902825 50 mg 50mg
17923495 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943825 Supplier Toronto Research Chemicals Supplier No. TRCP2183501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70458-92-3
Molecular Formula C17 H20 F N3 O3
Molecular Weight (g/mol) 333.36
Synonym 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo- (9CI, ACI), 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid (ACI), 1-Ethyl-6-fluoro-7-(4-methyl-piperazin-1-yl)-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid, 1584RB, 1589RB, Abactal, Abaktal, AM 725, EU 5306, Globacin, N'-Methylnorfloxacin, Pefbid, Peflacine, Pefloxacin, Pefloxacine, Pelox, Perflacin, Perti, 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 1584RB, 1589RB, AM 725, Abactal, Abaktal, EU 5306, Globacin, N'-Methylnorfloxacin, Pefbid, Peflacine, Pefloxacin, Pefloxacine, Pelox, Perflacin, Perti
IUPAC Name 1-ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H20FN3O3/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21/h8-10H,3-7H2,1-2H3,(H,23,24)
Formula Weight 333.1489
Percent Purity >95%
Chemical Name or Material Pefloxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.