Learn More
Thermo Scientific™ PEEKsil™ Capillary Tubing
Product Code. 11503920 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
0.002 in. ID; 10 cm length
0.002 in. ID; 20 cm length
0.002 in. ID; 50 cm length
0.004 in. ID; 10 cm length
0.004 in. ID; 20 cm length
0.004 in. ID; 50 cm length
0.007 in. ID; 10 cm length
0.007 in. ID; 20 cm length
0.007 in. ID; 50 cm length
Unit Size:
Pack of 2
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
11503920 0.002 in. ID; 10 cm length Pack of 5
10733697 0.002 in. ID; 20 cm length Pack of 5
11523920 0.002 in. ID; 50 cm length Pack of 2
10699485 0.004 in. ID; 10 cm length Pack of 5
10535475 0.004 in. ID; 20 cm length Pack of 5
11533920 0.004 in. ID; 50 cm length Pack of 2
10690426 0.007 in. ID; 10 cm length Pack of 5
10016654 0.007 in. ID; 20 cm length Pack of 5
11553920 0.007 in. ID; 50 cm length Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11503920 Supplier Thermo Scientific™ Supplier No. 60182500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get excellent chemical compatibility and very low carryover using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing.
Get excellent chemical compatibility and very low carryover to improve liquid chromatography (LC) performance using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing. Made of precision-bore fused silica tubing coated with a 1/16 in. O.D. PEEK covering, it is capable of withstanding high pressures. PEEKsil tubing is ideal for capillary HPLC and LC-mass spectrometry (MS). The very smooth internal surface ensures excellent flow characteristics and very low carryover. It is compatible with most organic solvents and is usable in most standard chromatography systems.
Recommended Use
  • Tubing is stiff; not recommended for uses requiring that tubing bends
  • Use precut lengths only; tube cutting after receipt may damage tubing
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC, LC/MS
Type Capillary Tubing
Description 0.002 in. ID; 10 cm length
For Use With (Application) HPLC, LC/MS
Diameter (English) Inner 0.002 in.
Diameter (English) Outer 1/16 in.
Diameter (Metric) Inner 0.051 mm
Diameter (Metric) Outer 1.588 mm
Overall Length (English) 3.937 in.
Overall Length (Metric) 10 cm
Material PEEK, Silica