Learn More
Cytiva PEEK Union Finger tight
Product Code. 11789555 Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
0.3 mm
Unit Size:
Pack of 4
Product Code. 11789555
Brand: Cytiva 11000852
Only null left
Add to basket
Add to basket
Union attachment. Cytiva PEEK Union Finger tight has an internal diameter of 0.3mm.
  • Packsize 4 
  • Polyetheretherketone (PEEK) 
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.3 mm