Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03031949
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10000251
  • 238.50 EUR / Pack of 10
Estimated Shipment: 20-06-2024
Description
  • Acc. to DIN 12383
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
According to DIN 12383
Borosilicate Glass, Duran Glass
85 mm
250 mL
19/26 NS