Learn More
Fisherbrand™ PE Tank with Level Sensor
Product Code. 15479679 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
30 L
60 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15479679 30 L Each
15489679 60 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15479679
Holds 30 or 60L. Fisher Scientific™ PE Tank with Level Sensor is equipped with a CO2 scavenger to capture and remove carbon dioxide.
  • Level sensor indicates amount of liquid in tank
  • Tank vent filter includes CO2 scavenger to protect purified water from atmospheric degradation.
  • Available in 30L or 60L
Specifications
Capacity (Metric) 30 L
For Use With (Equipment) Accu500
Type Tank, PE
For Use With (Application) Water purification
Material Polyethylene