missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (RM0004-3G28) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
200.00 EUR - 523.00 EUR
Specifications
| Antigen | PDGFR alpha |
|---|---|
| Clone | RM0004-3G28 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:400-1:800, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242793
|
Novus Biologicals
NB110-61020-0.025MG |
0.025 mg |
200.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078565
|
Novus Biologicals
NB110-61020 |
0.1 mg |
523.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PDGFR alpha | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Mouse | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| PDGFRA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| RM0004-3G28 | |
| Western Blot 1:400-1:800, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P26618 | |
| 5156 | |
| IgG2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title