missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
310.00 EUR - 504.00 EUR
Specifications
| Antigen | PDGFR alpha |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Research Discipline | Phospho Specific, Protein Kinase |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653250
|
Novus Biologicals
NBP2-76508-25ul |
25 μL |
310.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18372901
|
Novus Biologicals
NBP2-76508 |
100 μL |
504.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFR alpha Polyclonal specifically detects PDGFR alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PDGFR alpha | |
| Unconjugated | |
| Phospho Specific, Protein Kinase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 5156.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EVSWQYPMSEEESSDVEIRNEENNSGLFVTVLEVSSASAAHTGLYTCYYNHTQTEENELEGRHIYIYVPDPDVAFVPLGMTDYLVIVEDDDSAIIPCRTTDPETPVTLHNSEGVVPASYDSRQGFNG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Rabbit | |
| Human | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| PDGFRA | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title