Learn More
PDE1B Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503319
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| PDE1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q01064 | |
| PDE1B | |
| Full length human recombit protein of human PDE1B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E4 | |
| Unconjugated | |
| PDE1B | |
| HEL-S-79p, PDE1B1, PDES1B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5153 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |