missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE10A Polyclonal Antibody
GREENER_CHOICE

Product Code. 13465807
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
13465807 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 13465807

Brand: Invitrogen™ PA531293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, mouse brain, rat brain, Myc-DDK-tagged PDE10A-transfected 293T. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE10A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PDE10A
Gene Accession No. Q8CA95, Q9QYJ6, Q9Y233
Gene Alias ADSD2; cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); dual-substrate cAMP and cGMP, cyclic nucleotide phosphodiesterase; FLJ11894; FLJ25677; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19; PDE10A2 brain-specific isoform; Pde10a3; phosphodiesterase 10A; phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7); testis-specific phosphodiesterase PDE10A2
Gene Symbols PDE10A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PDE10A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10846, 23984, 63885
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.