Learn More
PDCL3 Monoclonal Antibody (OTI8D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809677S
Encompass_Preferred
 View more versions of this product
Product Code. 17562144
  • 242.00 EUR / 30µL
Description
PDCL3 Monoclonal antibody specifically detects PDCL3 in Human samples. It is validated for Western Blot
Specifications
Specifications
PDCL3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q99081
Mouse
Affinity chromatography
RUO
6938
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI8D6
Unconjugated
TCF12
1110061A19Rik; C80025; HTPHLP; IAP-associated factor VIAF1; PDCL3; PhLP2A; PHLP3; phosducin like 3; phosducin-like 3; phosducin-like protein 3; PhPL3; VIAF; VIAF1; VIAF-1; Viral IAP-associated factor 1
Full length human recombinant protein of human TCF12 produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions