Learn More
PDCL3 Monoclonal Antibody (OTI8D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809677S
This item is not returnable.
View return policy
Description
PDCL3 Monoclonal antibody specifically detects PDCL3 in Human samples. It is validated for Western BlotSpecifications
| PDCL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99081 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6938 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI8D6 | |
| Unconjugated | |
| TCF12 | |
| 1110061A19Rik; C80025; HTPHLP; IAP-associated factor VIAF1; PDCL3; PhLP2A; PHLP3; phosducin like 3; phosducin-like 3; phosducin-like protein 3; PhPL3; VIAF; VIAF1; VIAF-1; Viral IAP-associated factor 1 | |
| Full length human recombinant protein of human TCF12 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |