Learn More
Invitrogen™ PDCD6IP Monoclonal Antibody (14D10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA536229
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human Jurkat whole cell, human PANC-1 whole cell, human K562 whole cell, human SW579 whole cell, rat RH35 whole cell, mouse NIH3T3 whole cell. IHC: human mammary cancer tissue, human intestinal cancer tissue, human intestinal cancer tissue, mouse testis tissue, rat testis tissue, rat testis tissue. ICC/IF: U20S cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly.
Specifications
| PDCD6IP | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8WUM4, Q9QZA2, Q9WU78 | |
| Pdcd6ip | |
| E.coli-derived human PDCD6IP recombinant protein (Position: A2-D330). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 14D10 | |
| Unconjugated | |
| Pdcd6ip | |
| AI480591; AIP1; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; Alg2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein X; Alix; apoptosis-linked gene 2-interacting protein X; AW544830; C76364; dopamine receptor interacting protein 4; DRIP4; E2F1-inducible protein; Eig2; Hp95; KIAA1375; MGC17003; mKIAA1375; PDCD6-interacting protein; Pdcd6ip; pdcd6ip.L; programmed cell death 6 interacting protein; programmed cell death 6 interacting protein L homeolog; programmed cell death 6-interacting protein; putative signal tranduction protein Xp95; RGD1561176; Signal transduction protein Xp95; XELAEV_18031317mg; xp95 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 10015, 18571, 501083 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.