missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD173955, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
PD173955 is src family-selective tyrosine kinase inhibitor with IC50 of ∼22 nM for Src, Yes and Abl kinase
Specifications
Specifications
| Chemical Name or Material | PD173955 |
| Physical Form | Solid |
| CAS | 260415-63-2 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H16Cl2N4OS |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 10 mg/mL (22.56 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C1C(C2=C(Cl)C=CC=C2Cl)=CC3=CN=C(NC4=CC=CC(SC)=C4)N=C3N1C |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction