Learn More
PD 90780, MedChemExpress
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
PD 90780 is a non peptide antagonist of nerve growth factor (NGF) binding to the P75 NGF receptor, inhibits NGF-p75 NTR interaction with IC50s values of 23.1 μM and 1.8 μM in PC12 cells and PC12 nnr5 cells, respectively .
Specifications
Specifications
| Chemical Name or Material | PD 90780 |
| Physical Form | Solid |
| CAS | 77422-99-2 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H14N4O4 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 31.25 mg/mL (86.24 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | CN1C2=CC(C(O)=O)=NN2C(C3=CC(NC(C4=CC=CC=C4)=O)=CC=C13)=O |
| Show More |