missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCI 29732, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
PCI 29732 is a potent, orally active, reversible BTK inhibitor with Kiapp values of 8.2, 4.6, and 2.5 nM for BTK, Lck and Lyn, respectively. PCI 29732 shows only modest inhibitory activity against Itk, another Tec family kinase. PCI 29732 inhibits the function of ABCG2 by competitively binding to the ATP-binding site of ABCG2.
Specifications
Specifications
| Chemical Name or Material | PCI 29732 |
| CAS | 330786-25-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H21N5O |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 53 mg/mL (142.69 mM) ∣Ethanol : 10 mg/mL (26.92 mM; Need ultrasonic) |
| SMILES | NC1=NC=NC2=C1C(C3=CC=C(OC4=CC=CC=C4)C=C3)=NN2C5CCCC5 |
| Molecular Weight (g/mol) | 371.44 |
| Formula Weight | 371.44 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction