Learn More
MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Tablets
200 Tablets
500 Tablets
Unit Size:
100 tablets
200 tablets
500 tablets
Description
- No need for weighing and mixing chemicals
- Simply dissolve PBS tablets in water
- One tablet dissolved in 100mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
- Research use only
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Physical Form | Tablet |
| Quantity | 500 Tablets |
| Molecular Formula | ClK |
| MDL Number | MFCD00011360 |
| Includes | No Calcium, No Magnesium |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| SMILES | [Cl-].[K+] |
| IUPAC Name | potassium chloride |
| Molecular Weight (g/mol) | 74.55 |