missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure

Product Code. p-7082823 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
5kg
This item is not returnable. View return policy

Product Code. 15875168

Brand: Thermo Scientific J18280.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2 S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Packaging Plastic Bottle
Beilstein 1721722
Density 1.293
Quantity 100 g
Formula Weight 131.17
Percent Purity 98.5 to 101.5%
Specific Gravity 1.293
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Leucine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.