missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS SMO(W535L) MEM-EA Pharmacotrafficking Assay
Description
The PathHunter™ U2OS SMO(W535L) MEM-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting SMO. This cell line enables assessment of chaperone-based stabilization of SMO(W535L) via detection of rescued membrane trafficking. This product uses EFC technology. The included cell line overexpresses PK-tagged mutant SMO(W535L), which is misfolded and retained in the ER. The cell line also expresses membrane-localized EA. When a pharmacochaperone stabilizes the mutant SMO, it is trafficked to the membrane, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Pharmacotrafficking |
| Cell Line | U2OS |
| Format | 2 vials |
| For Use With (Application) | Cell Lines |
| Primary Agonist | SANT-1 |
| Quantity | 2 Vials |
| Target Entry | SMO(W535L) |
| Accession Number | NM_005631.4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?