missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Path-ID™ qPCR Master Mix with Manual
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
Description
Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
- AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
- Optimized buffer and dNTP concentration, for reliable PCR
- Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | 2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature. |
| Detection Method | Primer-probe |
| Form | Liquid |
| Format | Tube |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Fast or Standard |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | Ambion, Path-ID |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction