Learn More
Memmert Pass-through Oven UF450TS TwinDISPLAY, 449L, 20-250°C, Forced Convection
Brand:  Memmert UF450TS
Pass-through Oven UF450TS TwinDISPLAY, 449L, 20-250°C, Forced Convection
Product Code. 16308037
  • ONLINE EXCLUSIVE PRICE
    14050.00 EUR
    13624.00 EUR / Each
    Save 426.00 EUR
    3% Off
Only null left
Estimated Shipment: 06-02-2026