missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pasireotide L-aspartate salt, MedChemExpress
Description
Pasireotide (SOM230) L-aspartate salt, a long-acting cyclohexapeptide somatostatin analogue, can improve agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5, pKi=8.2/9.0/9.1/<7.0/9.9, respectively). Pasireotide L-aspartate salt exhibits antisecretory, antiproliferative, and proapoptotic activity.
Specifications
Specifications
| Chemical Name or Material | Pasireotide L-aspartate salt |
| CAS | 396091-77-3 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C62H73N11O13 |
| Quantity | 1 mg |
| Synonym | SOM230 L-aspartate |
| Solubility Information | DMSO : 1 mg/mL (0.85 mM; Need ultrasonic) |
| SMILES | N[C@@H](CC(O)=O)C(O)=O.NCCCC[C@@H](C(N[C@H]1CC2=CC=C(C=C2)OCC3=CC=CC=C3)=O)NC([C@H](NC([C@H](C4=CC=CC=C4)NC([C@H]5N(C([C@H](CC6=CC=CC=C6)NC1=O)=O)C[C@H](OC(NCCN)=O)C5)=O)=O)CC7=CNC8=C7C=CC=C8)=O |
| Molecular Weight (g/mol) | 1180.31 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?