Learn More
Pasiniazid, MedChemExpress
Product Code. 16422309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16422309 Supplier Medchem Express Supplier No. HYB1048
MedChemExpress Pasiniazid is an anti-TB and anti-leprosy drug, used to treat various types of TB and leprosy.
Pasiniazid is an anti-TB and anti-leprosy drug, used to treat various types of TB and leprosy.
Specifications
Chemical Name or Material Pasiniazid
CAS 2066-89-9
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C13H14N4O4
Quantity 1 g
Synonym Paraniazide Pasiniazide Isonicotinic acid hydrazide p-aminosalicylate
Solubility Information DMSO : ≥ 160 mg/mL (551.21 mM)
SMILES O=C(C1=CC=NC=C1)NN.O=C(O)C2=CC=C(N)C=C2O
Molecular Weight (g/mol) 290.27
Formula Weight 290.27
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation