Learn More
Brannan™ Partial Immersion PVC Coated Thermometer
Product Code. 12329359
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
Unit Size:
Each
Product Code. 12329359
Brand: Brannan™ 44/425/0
Only null left
Add to basket
Add to basket
Brannan™ Partial Immersion PVC Coated Thermometer is encapsulated in PVC to contain any glass or filling material in the event of breakage.
  • The breaksafe range of laboratory thermometers are encapsulated in PVC to contain any glass or filling material in the event of breakage
  • General purpose/stirring thermometer with black graduations
Specifications
Bottle Fill Green Spirit
Immersion Type Partial Immersion
Length (Metric) 200 mm
Material Glass
Product Type Partial Immersion Thermometer, PVC Coated
Max. Temperature (English) 230°F
Max. Temperature (Metric) 110°C
Min. Temperature (English) 14°F
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +110°C
Accuracy ±1.5°C
Subdivision 1°C
Immersion Length 76 mm
Width (Metric) 8 mm