missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARL Monoclonal Antibody (8C4B2;1H12E10)

Description
MA5-15428 targets PARL in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15428 immunogen is purified recombinant fragment of PARL (aa112-167) expressed in E. Coli..
This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | PARL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8C4B2;1H12E10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | PARL |
| Gene Accession No. | Q9H300 |
| Gene Alias | D16Ertd607e; Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Parl; parl {ECO:0000250; Pbeta; P-beta; presenilin associated rhomboid like; presenilin associated, rhomboid-like; presenilin associated, rhomboid-like isoform 1 preproprotein; presenilin associated, rhomboid-like preproprotein; presenilins-associated rhomboid-like protein, mitochondrial; PRO2207; Psarl; PSARL1; PSENIP2; RHBDS1; rhomboid 7 homolog 1; UniProtKB:Q9H300} |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?