Learn More
Paricalcitol, MedChemExpress
Product Code. 16481949
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481949 1 mg 1mg
16480598 5 mg 5mg
16448148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16481949 Supplier Medchem Express Supplier No. HY50919
MedChemExpress Paricalcitol, a vitamin D analogue, is a vitamin D receptor agonist, used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.
Paricalcitol, a vitamin D analogue, is a vitamin D receptor agonist, used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.
Specifications
Chemical Name or Material Paricalcitol
CAS 131918-61-1
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (240.02 mM; Need ultrasonic) ∣Ethanol : 12.5 mg/mL (30.00 mM; Need ultrasonic)
SMILES C[C@]([C@]([C@H](C)/C=C/[C@H](C)C(C)(C)O)([H])CC1)(CCC/2)[C@]1([H])C2=C\C=C3C[C@@H](O)C[C@H](O)C/3
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation