Learn More
Sartorius™ Grade LabSorb Polyethylene-Coated Papers
Product Code. 10150851 Shop All Sartorius Products
Click to view available options
:
Papers
Product Code. 10150851
Brand: Sartorius™ FT1601600050
Highly absorptive grade of paper coated on one side with polyethylene.
  • Surface protection paper
  • With cellulose side up, the paper absorbs liquids, which are stopped by the polyethylene layer preventing from soaking through
  • With polyethylene side up, useful for recovery of valuable or toxic liquids
TRUSTED_SUSTAINABILITY
Specifications
Color White