Learn More
Panidazole, MedChemExpress
Product Code. 16456247
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16456247
Brand: Medchem Express HY101715
MedChemExpress Panidazole is an amoebicide.
Panidazole is an amoebicide.
Specifications
Chemical Name or Material Panidazole
Physical Form Solid
CAS 13752-33-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H12N4O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 125 mg/mL (538.24 mM)
SMILES O=[N+](C1=CN=C(C)N1CCC2=CC=NC=C2)[O-]
Molecular Weight (g/mol) 232.24
Formula Weight 232.24
Percent Purity 98.77%
Grade Research