missing translation for 'onlineSavingsMsg'
Learn More

Pamiparib, MedChemExpress

Product Code. p-200097220
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493938

Brand: Medchem Express HY104044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pamiparib (BGB-290) is an orally active, potent, highly selective PARP inhibitor, with IC50 values of 0.9 nM and 0.5 nM for PARP1 and PARP2, respectively. Pamiparib has potent PARP trapping, and capability to penetrate the brain, and can be used for the research of various cancers including the solid tumor.

Pamiparib (BGB-290) is an orally active, potent, highly selective PARP inhibitor, with IC50 values of 0.9 nM and 0.5 nM for PARP1 and PARP2, respectively. Pamiparib has potent PARP trapping, and capability to penetrate the brain, and can be used for the research of various cancers including the solid tumor.

Specifications

Chemical Name or Material Pamiparib
CAS 1446261-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H15FN4O
Quantity 10 mg
Synonym BGB-290
Solubility Information DMSO : 62.5 mg/mL (209.51 mM; Need ultrasonic)
SMILES O=C1NN=C2CN(CCC3)[C@@]3(C)C(N4)=C2C5=C4C=C(F)C=C51
Molecular Weight (g/mol) 298.31
Formula Weight 298.31
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.