missing translation for 'onlineSavingsMsg'
Learn More

Pamidronic Acid Sodium Salt Hydrate, TRC

Product Code. 30420605
Change view
Click to view available options
Quantity:
50 mg
100 mg
50 g
Unit Size:
100mg
50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420605 100 mg 100mg
30409016 50 mg 50mg
30425068 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420605 Supplier Toronto Research Chemicals Supplier No. TRCP172500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pamidronic Acid Sodium Salt Hydrate
Molecular Formula C3 H9 N O7 P2 . 2 Na . xH2O
Quantity 100 mg
InChI Formula InChI=1S/C3H11NO7P2.2Na.H2O.2H/c4-2-1-3(5,12(6,7)8)13(9,10)11;;;;;/h5H,1-2,4H2,(H2,6,7,8)(H2,9,10,11);;;1H2;;
SMILES O.[Na].[Na].NCCC(O)(P(=O)(O)O)P(=O)(O)O
IUPAC Name (3-amino-1-hydroxy-1-phosphono-propyl)phosphonic acid;sodium hydride;hydrate
Molecular Weight (g/mol) 235.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.