missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PALMD Polyclonal Antibody
GREENER_CHOICE

Product Code. 15781611
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15781611 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15781611 Supplier Invitrogen™ Supplier No. PA553678

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YDDGQKSVYA VSSNHSAAYN GTDGLAPVEV EELLRQASER NSKSPTEYHE PVYANPFYRP TTPQRETVTP GPNFQERIKI KTNGLGIGVN ESIHNMGNGL SEERGNNFNH ISPIPPVPHP RSVIQQAEEK LHTP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.

PALMD gene ontology annotations related to this gene include regulation of cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PALMD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PALMD
Gene Accession No. Q9NP74
Gene Alias 4631423C22Rik; C1orf11; PALMD; Palmdelphin; PALML; Paralemmin-like protein; paralemmin-related; paralemnin-like protein
Gene Symbols PALMD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PALMD. Recombinant protein control fragment (Product #RP-90708).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.