Learn More
Sampling Systems™ Pallet Knives with Wooden Handles
Product Code. 15869591 Shop All Sampling Systems Products
Click to view available options
Length (Metric) Blade:
100 mm
200 mm
300 mm
Width (Metric) Blade:
20 mm
30 mm
35 mm
Unit Size:
Each
Product Code. 15869591
Brand: Sampling Systems™ A201100
Only null left
Add to basket
Add to basket
These Pallet Knives are excellent for general sampling purposes in the laboratory.
  • Stainless steel blade with wooden handle
  • Satin finish
  • Range of sizes available
Specifications
Product Type Pallet Knife
Length (Metric) Blade 100 mm
Width (Metric) Blade 20 mm
Material Stainless Steel, Wood
Length (Metric) 190 mm