missing translation for 'onlineSavingsMsg'
Learn More

Palladium on activated carbon, 3% Pd, unreduced, Thermo Scientific Chemicals

Product Code. p-42583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10687182 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687182 Supplier Thermo Scientific Acros Supplier No. 217870100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3 , 7440-44-0
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Black
Quantity 10 g
Packaging Glass bottle
Particle Size Typical particle size distribution: D50: 20 Micron, < 8 Micron: 17%, <16 Micron: 39%, <32 Micron: 67%, <64Micron: 90%, <128 Micron: 99%
Form Powder
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Formula Weight 106.42
Percent Purity 3% (Pd)
Chemical Name or Material Palladium on activated carbon, 3% Pd
compliance-icons
Product Identifier
  • Palladium on activated carbon, unreduced
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P233-Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.